missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrothiophene 99.0+%, TCI America™

Catalog No. T0114500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0114500ML 500 mL
T011425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0114500ML Supplier TCI America Supplier No. T0114500ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 110-01-0
Molecular Formula C4H8S
Molecular Weight (g/mol) 88.17
MDL Number MFCD00005476
InChI Key RAOIDOHSFRTOEL-UHFFFAOYSA-N
Synonym tetrahydrothiophene, thiophene, tetrahydro, thiophane, thiacyclopentane, tetramethylene sulfide, thilane, tetrahydrothiophen, pennodorant 1013, thiofan, thiolan
PubChem CID 1127
ChEBI CHEBI:48458
IUPAC Name thiolane
SMILES C1CCSC1

Specifications

Color Colorless
Boiling Point 121°C
Quantity 500 mL
UN Number 2412
Formula Weight 88.17
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetrahydrothiophene
EINECSNumber (5)-0075
RTECSNumber XN0370000
TSCA Yes