missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrothiophene 1,1-Dioxide 99.0+%, TCI America™

Catalog No. T0115500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0115500G 500 g
T011525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0115500G Supplier TCI America Supplier No. T0115500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 126-33-0
Molecular Formula C4H8O2S
Molecular Weight (g/mol) 120.17
MDL Number MFCD00005484
InChI Key HXJUTPCZVOIRIF-UHFFFAOYSA-N
Synonym sulfolane, tetramethylene sulfone, tetrahydrothiophene 1,1-dioxide, sulfolan, sulpholane, sulfalone, sulphoxaline, 1,1-dioxothiolan, dioxothiolan, thiophan sulfone
PubChem CID 31347
ChEBI CHEBI:74794
IUPAC Name 1λ⁶-thiolane-1,1-dione
SMILES O=S1(=O)CCCC1

Specifications

Color White-Yellow
Boiling Point 285°C
Quantity 500 g
Formula Weight 120.17
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Tetrahydrothiophene 1,1-Dioxide
EINECSNumber (5)-0077
RTECSNumber XN0700000
TSCA Yes