missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydropyran-4-methanol 97.0+%, TCI America™

Catalog No. T29651G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T29651G Supplier TCI America Supplier No. T29651G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14774-37-9
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00457804
InChI Key YSNVSVCWTBLLRW-UHFFFAOYSA-N
Synonym tetrahydropyran-4-methanol, tetrahydro-2h-pyran-4-yl methanol, 4-hydroxymethyl tetrahydropyran, oxan-4-yl methanol, tetrahydro-pyran-4-yl-methanol, tetrahydro-2h-pyran-4-ylmethanol, 2h-pyran-4-methanol, tetrahydro, 4-hydroxymethyltetrahydropyran, oxan-4-yl-methanol
PubChem CID 2773573
IUPAC Name (oxan-4-yl)methanol
SMILES OCC1CCOCC1

Specifications

Color Yellow
Boiling Point 110°C
Quantity 1 g
Formula Weight 116.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetrahydropyran-4-methanol