missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran/Propylene Glycol, 80/20 v/v, Reagents

Catalog No. 01333451
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333451 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01333451 Supplier Labchem Inc Supplier No. CT1005004C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tetrahydrofuran/Propylene Glycol, 80/20 v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 109-99-9 , 57-55-6
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym Oxolane/1,2-Propanediol
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Color Colorless
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.9186
Grade Reagent
Chemical Name or Material Tetrahydrofuran/Propylene Glycol, 80/20 v/v