missing translation for 'onlineSavingsMsg'
Learn More

Tétrahydro-2H-thiopyran-4-ol, 97%, Thermo Scientific™

Numéro de catalogue. p-3830419
Modifier l'affichage
Click to view available options
Quantité:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC61915DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC61915DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC61915DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 29683-23-6
Formule moléculaire C5H10OS
Poids moléculaire (g/mol) 118.19
Numéro MDL MFCD02942489
Clé InChI YODQQARABJQLIP-UHFFFAOYSA-N
Synonyme tetrahydro-2h-thiopyran-4-ol, tetrahydrothiopyran-4-ol, thiacyclohexane-4-ol, 2h-thiopyran-4-ol, tetrahydro, 4-hydroxythiane, acmc-209ydy, tetrahydrothiapyran-4-ol, 4-hydroxytetrahydrothiopyran, 4-hydroxy-tetrahydrothiopyran, tetrahydro-4h-thiopyran-4-ol
PubChem CID 576296
Nom de l’IUPAC thian-4-ol
SOURIRES OC1CCSCC1

Spécifications

Nom ou substance chimique Tetrahydro-2H-thiopyran-4-ol
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Bouteille en verre ambrée
Quantité 1 g
Poids de formule 118.2
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.