missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-2H-pyran-4-carbonyl chloride, ≥97%, Thermo Scientific™

Catalog No. CC29902DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC29902DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC29902DA Supplier Thermo Scientific Chemicals Supplier No. CC29902DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 40191-32-0
Molecular Formula C6H9ClO2
Molecular Weight (g/mol) 148.59
MDL Number MFCD06200863
InChI Key RYGUCYSSMOFTSH-UHFFFAOYSA-N
Synonym tetrahydro-2h-pyran-4-carbonyl chloride, 2h-pyran-4-carbonyl chloride, tetrahydro, tetrahydro-2h-pyran-4-carbonylchloride, pubchem12437, ksc235m5r, tetrahydropyran-4-carbonyl chloride, tetrahydro-pyran-4-carbonyl chloride, 4-chloroformyl tetrahydro-2h-pyran, tetrahydropyran-4-carboxylic chloride, 4-chlorocarbonyl tetrahydro-2h-pyran
PubChem CID 2795505
IUPAC Name oxane-4-carbonyl chloride
SMILES ClC(=O)C1CCOCC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 148.59
Percent Purity 95%
Chemical Name or Material Tetrahydro-2H-pyran-4-carbonyl chloride
Hazard Category Danger
Hazard Statement GHS P Statement Causes severe skin burns and eye damage. Reacts violently with water. Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair):