missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahexylammonium Hydroxide (10% in Methanol), TCI America™
Supplier: TCI America T11345G
Specifications
Tetrahexylammonium Hydroxide (10% in Methanol) | |
Yellow | |
MFCD00059255 | |
2924 | |
CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC.[OH-] | |
371.694 | |
371.69 |
17756-56-8 | |
C24H53NO | |
5 g | |
JCJNUSDBRRKQPC-UHFFFAOYSA-M | |
tetrahexylazanium;hydroxide | |
13998878 | |
Liquid |
Chemical Identifiers
17756-56-8 | |
371.694 | |
JCJNUSDBRRKQPC-UHFFFAOYSA-M | |
tetrahexylazanium;hydroxide |
C24H53NO | |
MFCD00059255 | |
13998878 | |
CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC.[OH-] |
Safety and Handling
EINECSNumber : (2)-0186
TSCA : No