missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylthiuram Disulfide, 97%

Catalog No. AC138125000
Change view
Click to view available options
Quantity:
25 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC138125000 500 g Plastic bottle
AC138120250 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC138125000 Supplier Thermo Scientific Chemicals Supplier No. 138125000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 97-77-8 | C10H20N2S4 | 296.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97-77-8
Molecular Formula C10H20N2S4
Molecular Weight (g/mol) 296.51
MDL Number MFCD00008157
InChI Key AUZONCFQVSMFAP-UHFFFAOYSA-N
Synonym disulfiram, tetraethylthiuram disulfide, antabuse, alcophobin, antabus, anticol, bis diethylthiocarbamoyl disulfide, esperal, teturam, tetd
PubChem CID 3117
ChEBI CHEBI:4659
IUPAC Name diethylcarbamothioylsulfanyl N,N-diethylcarbamodithioate
SMILES CCN(CC)C(=S)SSC(=S)N(CC)CC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 70.0°C to 74.0°C
Color Yellow
Boiling Point 117°C (23.0 hPa)
Loss on Drying 0.3% max. (60°C)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Linear Formula [(C2H5)2NCS2-]2
Quantity 500 g
Beilstein 04, 122
Fieser 06,569
Merck Index 15, 3407
Solubility Information Solubility in water: 0.02g/100mL. Other solubilities: 3.82g/100mL alcohol, 7.14g/100mL ether, soluble in acetone, benzene, carbon disulfice and, chloroform
Formula Weight 296.51
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Granules
Chemical Name or Material Tetraethylthiuram disulfide, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-607-8
RTECSNumber JO1225000
TSCA TSCA

RUO – Research Use Only