missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethylsilane, 97%, Thermo Scientific™
Spécifications
Spécifications
| Nom ou substance chimique | Tetraethylsilane |
| Densité | 0.7600g/mL |
| Point d’ébullition | 153.0°C to 154.0°C |
| Point d’éclair | 26°C |
| Spectre infrarouge | Authentic |
| Fourchette de pourcentages d’essai | 96% min. (GC) |
| Emballage | Glass bottle |
| Indice de réfraction | 1.4250 to 1.4280 |
| Formule linéaire | Si(C2H5)4 |
| Quantité | 5g |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and ea |
| EINECSNumber | 211-155- |
RUO – Research Use Only