missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylgermane 98.0+%, TCI America™

Catalog No. T11571G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T11571G Supplier TCI America Supplier No. T11571G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 597-63-7
Molecular Formula C8H20Ge
Molecular Weight (g/mol) 188.878
MDL Number MFCD00015096
InChI Key QQXSEZVCKAEYQJ-UHFFFAOYSA-N
Synonym tetraethylgermanium, germane, tetraethyl, tetraethyl germane, germanium, tetraethyl, acmc-209mfg, c2h5 4ge, qqxsezvckaeyqj-uhfffaoysa, wln: 2-ge-2&2&2
PubChem CID 11703
IUPAC Name tetraethylgermane
SMILES CC[Ge](CC)(CC)CC

Specifications

Color Colorless
Boiling Point 165°C
Quantity 1 g
UN Number 1993
Formula Weight 188.88
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetraethylgermane
RTECSNumber LY5290000
TSCA Yes