missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylenepentamine (so called), TCI America™

Catalog No. T009825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T009825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T009825G Supplier TCI America Supplier No. T009825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112-57-2
Molecular Formula C8H23N5
Molecular Weight (g/mol) 189.31
MDL Number MFCD00008168
InChI Key FAGUFWYHJQFNRV-UHFFFAOYSA-N
Synonym tetraethylenepentamine, tetren, 1,4,7,10,13-pentaazatridecane, tetraethylene pentamine, tetraethylpentylamine, 1,11-diamino-3,6,9-triazaundecane, 3,6,9-triazaundecamethylenediamine, 3,6,9-triazaundecane-1,11-diamine, deh 26, unii-yzd1c9kq28
PubChem CID 8197
ChEBI CHEBI:49798
IUPAC Name (2-aminoethyl)[2-({2-[(2-aminoethyl)amino]ethyl}amino)ethyl]amine
SMILES NCCNCCNCCNCCN

Specifications

Melting Point -40°C
Color Yellow
Boiling Point 375°C
Quantity 25 g
UN Number 2320
Formula Weight 189.31
Physical Form Liquid
Chemical Name or Material Tetraethylenepentamine (so called)
EINECSNumber (2)-0162
RTECSNumber KH8585000
TSCA Yes