missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylene Glycol 95.0+%, TCI America™

Catalog No. T009925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T009925G 25 g
T0099500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T009925G Supplier TCI America Supplier No. T009925G

Chemical Identifiers

CAS 112-60-7
Molecular Formula C8H18O5
Molecular Weight (g/mol) 194.227
MDL Number MFCD00002879
InChI Key UWHCKJMYHZGTIT-UHFFFAOYSA-N
Synonym tetraethylene glycol, tetraglycol, 2,2'-oxybis ethane-2,1-diyl bis oxy diethanol, hi-dry, carbitol, diethyl, 3,6,9-trioxaundecane-1,11-diol, bis 2-2-hydroxyethoxy ethyl ether, ethanol, 2,2'-oxybis 2,1-ethanediyloxy bis, tetra ethylene glycol, hsdb 843
PubChem CID 8200
ChEBI CHEBI:44920
IUPAC Name 2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethanol
SMILES C(COCCOCCOCCO)O

Specifications

Color Yellow
pH 6 to 7 (50% H2O soln.)
Boiling Point 328°C
Quantity 25 g
Formula Weight 194.23
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetraethylene Glycol
EINECSNumber (2)-0441
RTECSNumber XC2100000
TSCA Yes