missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium chloride

Catalog No. AAJ6250714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6250714 25 g
AAJ6250722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6250714 Supplier Thermo Scientific Chemicals Supplier No. J6250714
Only null left

Phase transfer catalyst | CAS: 56-34-8 | C8H20ClN | 165.71 g/mol

Tetraethylammonium chloride is used as a catalyst. Tetraethylammonium Chloride can be used as a source of tetraethylammonium ions for various pharmaceutical studies and has the ability to block K+ channels in various tissues. Tetraethylammonium Chloride can also block the transmission of nervous impulses across autonomic ganglions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethylammonium chloride is used as a catalyst. Tetraethylammonium Chloride can be used as a source of tetraethylammonium ions for various pharmaceutical studies and has the ability to block K+ channels in various tissues. Tetraethylammonium Chloride can also block the transmission of nervous impulses across autonomic ganglions.

Solubility
Soluble in water, ethanol, chloroform, or acetone

Notes
Hygroscopic, Incompatible with strong oxidizing agents. Protect from moisture.  Keep container tightly closed in a dry and well-ventilated place. Handle under argon. Store in cool place.

Chemical Identifiers

CAS 56-34-8
Molecular Formula C8H20ClN
Molecular Weight (g/mol) 165.71
MDL Number MFCD00149992
InChI Key YMBCJWGVCUEGHA-UHFFFAOYSA-M
Synonym tetraethylammonium chloride, tetraethyl ammonium chloride, etamon chloride, tea chloride, teac, n,n,n-triethylethanaminium chloride, tetraethylammoniumchloride, ethanaminium, n,n,n-triethyl-, chloride, tetraethyl-ammonium chloride, unii-8b82z83xnn
PubChem CID 5946
ChEBI CHEBI:78161
IUPAC Name tetraethylazanium;chloride
SMILES [Cl-].CC[N+](CC)(CC)CC

Specifications

Color White to Yellow
Linear Formula (CH3CH2)4NCl
Quantity 25 g
Beilstein 3563247
Sensitivity Hygroscopic
Merck Index 14,9200
Solubility Information Soluble in water,ethanol,chloroform,or acetone
Formula Weight 165.71
Physical Form Powder
Chemical Name or Material Tetraethylammonium chloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-267-5
RTECSNumber BS6125000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only