missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium bromide, 99+%

Catalog No. AC220715000
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC220715000 500 g Glass bottle
AC220710500 50 g Glass bottle
AC220712500 250 g Glass bottle
AC220710025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC220715000 Supplier Thermo Scientific Chemicals Supplier No. 220715000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 71-91-0 | C8H20BrN | 210.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst

Chemical Identifiers

CAS 71-91-0
Molecular Formula C8H20BrN
Molecular Weight (g/mol) 210.16
InChI Key HWCKGOZZJDHMNC-UHFFFAOYSA-M
Synonym tetraethylammonium bromide, teab, tea bromide, tetrylammonium bromide, sympatektoman, tetranium, etylon, bromethyl, beparon, etambro
PubChem CID 6285
IUPAC Name tetraethylazanium;bromide
SMILES CC[N+](CC)(CC)CC.[Br-]

Specifications

Melting Point 285.0°C
Color White to Cream
Loss on Drying 1.0% max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (Argentometry)
Packaging Glass bottle
Linear Formula (C2H5)4NBr
Quantity 500 g
Beilstein 04,104
Fieser 06,568; 13,46
Merck Index 15,9344
Solubility Information Solubility in water: 2795g/L (25°C). Other solubilities: freely soluble in alcohol, chloroform and acetone, slightly soluble in benzene
Formula Weight 210.16
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Tetraethylammonium bromide
Show More Show Less
EINECSNumber 200-769-4

RUO – Research Use Only