missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium Bromide, 40% (w/w) in Glacial Acetic Acid, Ricca Chemical

Catalog No. 838532
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
838532 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 838532 Supplier Ricca Chemical Company Supplier No. 838532

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 71-91-0 , 64-19-7
Molecular Formula C8H20BrN
Molecular Weight (g/mol) 210.159
InChI Key HWCKGOZZJDHMNC-UHFFFAOYSA-M
Synonym tetraethylammonium bromide, teab, tea bromide, tetrylammonium bromide, sympatektoman, tetranium, etylon, bromethyl, beparon, etambro
PubChem CID 6285
IUPAC Name tetraethylazanium;bromide
SMILES CC[N+](CC)(CC)CC.[Br-]

Specifications

Color Golden-Tan
CAS Min % 60.0, 38.07
CAS Max % 62.0, 39.63
Percent Purity 40%
Grade Laboratory
Packaging Amber Glass Bottle
Solvent or Matrix Glacial Acetic Acid
pH <2
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 40% (w/w)
Chemical Name or Material Tetraethylammonium Bromide
Show More Show Less