missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyl Orthotitanate (contains 35% Tetraisopropyl Orthotitanate at maximum), TCI America™

Catalog No. T096725ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T096725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T096725ML Supplier TCI America Supplier No. T096725ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3087-36-3
Molecular Formula C8H20O4Ti
Molecular Weight (g/mol) 228.111
MDL Number MFCD00009071
InChI Key JMXKSZRRTHPKDL-UHFFFAOYSA-N
Synonym titanium iv ethoxide, ethyl titanate, titanium tetraethoxide, titanium ethoxide, tetraethyl titanate, titanium iv ethanolate, titanium 4+ tetraethanolate, titanium 4+ ethanolate
PubChem CID 76524
IUPAC Name ethanolate;titanium(4+)
SMILES CC[O-].CC[O-].CC[O-].CC[O-].[Ti+4]

Specifications

Color Yellow
Boiling Point 236°C
Quantity 25 mL
UN Number 1993
Formula Weight 228.11
Physical Form Liquid
Chemical Name or Material Tetraethyl Orthotitanate (contains 35% Tetraisopropyl Orthotitanate at maximum)
EINECSNumber (2)-0228&(2)-2150
TSCA Yes