Learn More
Tetraethyl orthocarbonate, 97+%
Description
Tetraethyl Orthocarbonate is used in the synthesis of chemokine receptor-5 inhibitors against HIV-1, benzobisoxazoles and in the preparation of organic semiconductors. It is also employed in the synthesis of 2,7-dimethylene-1,4,6,9-tetraoxaspiro[4,4]nonane and in the synthesis of cross linked poly(orthocarbonate)s used as organic solvent absorbent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tetraethyl Orthocarbonate is used in the synthesis of chemokine receptor-5 inhibitors against HIV-1, benzobisoxazoles and in the preparation of organic semiconductors. It is also employed in the synthesis of 2,7-dimethylene-1,4,6,9-tetraoxaspiro[4,4]nonane and in the synthesis of cross linked poly(orthocarbonate)s used as organic solvent absorbent.
Solubility
Insoluble in water. Contact with water releases a flammable alcohol. Soluble in chloroform and methanol.
Notes
Moisture Sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Store under inert atmosphere and refrigerated.
Specifications
Specifications
| Density | 0.917 |
| Boiling Point | 158°C to 161°C |
| Flash Point | 52°C (125°F) |
| Refractive Index | 1.393 |
| Quantity | 100 g |
| UN Number | UN3272 |
| Beilstein | 1747401 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Insoluble in water. Contact with water releases a flammable alcohol. Soluble in chloroform and methanol. |
| Formula Weight | 192.26 |
| Show More |
Safety and Handling
| Hazard Category | H226-H315-H319-H335 |
| Hazard Statement | GHS H Statement H226 Flammable liquid and vapour. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S. |
| EINECSNumber | 201-082-2 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only