missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyl ethylenediphosphonate, 98%

Catalog No. AAA1420614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1420614 25 g
AAA1420603 1 g
AAA1420606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1420614 Supplier Thermo Scientific Chemicals Supplier No. A1420614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 995-32-4 | C10H24O6P2 | 302.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 995-32-4
Molecular Formula C10H24O6P2
Molecular Weight (g/mol) 302.24
MDL Number MFCD00015133
InChI Key RSQYXXACEZCDFS-UHFFFAOYSA-N
Synonym tetraethyl ethylenediphosphonate, tetraethyl ethylenebisphosphonate, ethylenediphosphonic acid tetraethyl ester, tetraethyl ethane-1,2-diylbis phosphonate, 1,2-bis diethoxyphosphoryl ethane, 1,2-bis diethyl-phosphonato ethane, phosphonic acid, 1,2-ethanediylbis-, tetraethyl ester, diethyl 2-diethoxyphosphoryl ethylphosphonate, phosphonic acid, ethylenedi-, tetraethyl ester, acmc-209scy
PubChem CID 70446
IUPAC Name 1,2-bis(diethoxyphosphoryl)ethane
SMILES CCOP(=O)(CCP(=O)(OCC)OCC)OCC

Specifications

Density 1.146
Boiling Point 153°C to 154°C (0.9 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.441
Quantity 25 g
Beilstein 1800096
Formula Weight 302.25
Percent Purity 98%
Chemical Name or Material Tetraethyl ethylenediphosphonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-625-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only