missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyl 4-tert-Butylcalix[4]arene-O,O',O″,O‴-tetraacetate 96.0+%, TCI America™

Catalog No. T2563200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2563200MG 200 mg
T25631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T2563200MG Supplier TCI America Supplier No. T2563200MG

Chemical Identifiers

CAS 97600-39-0
Molecular Formula C60H80O12
Molecular Weight (g/mol) 993.288
MDL Number MFCD00145373
InChI Key HZHADWCIBZZJNV-UHFFFAOYSA-N
Synonym sodium ionophore x, 4-tert-butylcalix 4 arene-tetraacetic acid tetraethyl ester, tetraethyl 4-tert-butylcalix 4 arene-o,o',o,o'-tetraacetate, p-tert-butylcalix 4 arene tetrakis ethyl acetate, sodium ionophore x, selectophore tm , function tested, 4-t-butylcalix 4 arene-tetraacetic acid tetraethyl ester, 4-tert-butylcalix 4 arene-tetraacetic acid tetraet, 4-tert-butylcalix 4 arenetetraacetic acid tetraethyl ester, 4-tert-butylcalix 4 arene-o,o',o,o'-tetraacetic acid tetraethyl ester
PubChem CID 2724889
SMILES CCOC(=O)COC1=C2CC3=CC(=CC(=C3OCC(=O)OCC)CC4=C(C(=CC(=C4)C(C)(C)C)CC5=C(C(=CC(=C5)C(C)(C)C)CC1=CC(=C2)C(C)(C)C)OCC(=O)OCC)OCC(=O)OCC)C(C)(C)C

Specifications

Melting Point 155°C
Color White-Yellow
Quantity 200 mg
Formula Weight 993.29
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Tetraethyl 4-tert-Butylcalix[4]arene-O,O′,O′′,O′′′-tetraacetate