missing translation for 'onlineSavingsMsg'
Learn More

Tetradecylbenzene 97.0+%, TCI America™

Catalog No. T08665G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T08665G 5 g
T086625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T08665G Supplier TCI America Supplier No. T08665G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1459-10-5
Molecular Formula C20H34
Molecular Weight (g/mol) 274.492
MDL Number MFCD00015080
InChI Key JZALLXAUNPOCEU-UHFFFAOYSA-N
Synonym 1-phenyltetradecane, benzene, tetradecyl, 1-tetradecylbenzene, tetradecane, 1-phenyl, myristylbenzene, n-tetradecylbenzene, unii-i0y9j0o95a, phenyl-n-tetradecane, acmc-209cvz, 1-phenyltetradecane, analytical standard
PubChem CID 15086
IUPAC Name tetradecylbenzene
SMILES CCCCCCCCCCCCCCC1=CC=CC=C1

Specifications

Melting Point 16°C
Color Yellow
Boiling Point 207°C
Quantity 5 g
Formula Weight 274.49
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetradecylbenzene
EINECSNumber (3)-0021
TSCA Yes