missing translation for 'onlineSavingsMsg'
Learn More

Tetradecylamine 96.0+%, TCI America™

Catalog No. T0090400G
Change view
Click to view available options
Quantity:
25 g
400 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0090400G 400 g
T009025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0090400G Supplier TCI America Supplier No. T0090400G

Chemical Identifiers

CAS 2016-42-4
Molecular Formula C14H31N
Molecular Weight (g/mol) 213.409
MDL Number MFCD00008157
InChI Key PLZVEHJLHYMBBY-UHFFFAOYSA-N
Synonym tetradecylamine, 1-tetradecylamine, myristylamine, 1-tetradecanamine, n-tetradecylamine, 1-aminotetradecane, tetradecanamine, armeen 14, monotetradecylamine, alamine 5d
PubChem CID 16217
IUPAC Name tetradecan-1-amine
SMILES CCCCCCCCCCCCCCN

Specifications

Color White-Yellow
Boiling Point 291°C
Quantity 400 g
UN Number 3259
Formula Weight 213.41
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Tetradecylamine
EINECSNumber (2)-0133&(2)-0176
RTECSNumber XB7970000
TSCA Yes