missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetradecyl sodium sulfate, 95%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Nom ou substance chimique | Tetradecyl sodium sulfate |
| Point de fusion | 199.0°C |
| Couleur | White |
| Analyse élémentaire | 94% min. |
| Spectre infrarouge | Authentic |
| Fourchette de pourcentages d’essai | 94% min |
| Emballage | Glass bottle |
| Formule linéaire | CH3(CH2)13OSO3Na |
| Quantité | 5 g |
| Poids de formule | 316.44 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. |
| EINECSNumber | 214-737-2 |
RUO – Research Use Only