missing translation for 'onlineSavingsMsg'
Learn More

Tetradecane (Reagent), Fisher Chemical

Catalog No. O4595500
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
O4595500 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O4595500 Supplier Thermo Fisher Scientific Supplier No. O4595 500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 629-59-4
Molecular Formula C14H30
Molecular Weight (g/mol) 198.394
MDL Number MFCD00008986
InChI Key BGHCVCJVXZWKCC-UHFFFAOYSA-N
Synonym n-tetradecane, alkanes, c14-16, tridecane, methyl, ccris 715, tetradecane, analytical standard, alkanes, c14-30, myristyl, tetradekan, n-teradecane, olefine
PubChem CID 12389
ChEBI CHEBI:41253
IUPAC Name tetradecane
SMILES CCCCCCCCCCCCCC

Specifications

Melting Point 5.8°C
Color Colorless
Density 0.767g/cm³
Boiling Point 252°C
Flash Point 99°C (210.2°F)
Melting Range 4° to 8°C
Assay Percent Range ≥98 %
Packaging Glass Bottle
Linear Formula CH3(CH2)12CH3
Quantity 500 mL
Formula Weight 198.39
Percent Purity ≥98%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Tetradecane
Show More Show Less