missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetracycline Hydrochloride USP , Medisca™
$147.19 - $2428.93
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
18-048-044
|
Medisca Pharmaceutique Inc
005305 |
100 g |
Each for $444.69
|
|
|||||
|
18-048-062
|
Medisca Pharmaceutique Inc
005308 |
500 g |
Each for $1,423.01
|
|
|||||
|
18-048-063
|
Medisca Pharmaceutique Inc
005309 |
1 kg |
Each for $2,428.93
|
|
|||||
|
18-048-123
|
Medisca Pharmaceutique Inc
005304 |
25 g |
Each for $147.19
|
|
|||||
Description
Tetracycline Hydrochloride USP is a yellow odorless crystalline powder. It is moderately hygroscopic. Stable in air, but exposure to strong sunlight in moist air causes it to darken.
Chemical Identifiers
| 64-75-5 |
Specifications
| α-Carboxybenzyl Penicillin Disodium Salt | |
| 64-75-5 | |
| 100 g | |
| USP |
| Crystalline Powder | |
| Yellow | |
| ≥ 900 μg/mg (C22H24N2O8 HCl) |