missing translation for 'onlineSavingsMsg'
Learn More

Tetracycline Hydrochloride USP , Medisca™
GREENER_CHOICE

Catalog No. 18048044
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18048044 100 g
18048062 500 g
18048063 1 kg
18048123 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18048044 Supplier Medisca Pharmaceutique Inc Supplier No. 005305

Tetracycline Hydrochloride USP is a yellow odorless crystalline powder. It is moderately hygroscopic. Stable in air, but exposure to strong sunlight in moist air causes it to darken.

Chemical Identifiers

CAS 64-75-5

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Crystalline Powder
Color Yellow
Quantity 100 g
Percent Purity ≥ 900 μg/mg (C22H24N2O8 HCl)
Grade USP