Learn More
Tetracycline Hydrochloride USP , Medisca™
Supplier: Medisca Pharmaceutique Inc 005309
Description
Tetracycline Hydrochloride USP is a yellow odorless crystalline powder. It is moderately hygroscopic. Stable in air, but exposure to strong sunlight in moist air causes it to darken.
Specifications
| α-Carboxybenzyl Penicillin Disodium Salt | |
| 64-75-5 | |
| 1 kg | |
| USP |
| Crystalline Powder | |
| Yellow | |
| ≥ 900 μg/mg (C22H24N2O8 HCl) |
Chemical Identifiers
| 64-75-5 |