missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. T156825G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T156825G 25 g
T156810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T156825G Supplier TCI America Supplier No. T156825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35895-70-6
Molecular Formula C18H36F3NO2
Molecular Weight (g/mol) 355.49
MDL Number MFCD00042585
InChI Key WTEXQPWIUJQYJQ-UHFFFAOYSA-M
Synonym tetrabutylammonium triflate, tetrabutylammonium trifluoromethanesulfonate, tetra-n-butylammonium trifluoromethanesulfonate, tetrabutylammonium ion triflate, trifluoromethanesulfonic acid tetrabutylammonium salt, acmc-1aglb, tetrakis but-1-yl ammonium triflate, tetrabutylazanium; trifluoromethanesulfonate, tetra-n-butylammonium trifluoromethanesulphonate
PubChem CID 2733306
IUPAC Name tetrabutylazanium trifluoroacetate
SMILES [O-]C(=O)C(F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 113°C
Color White-Yellow
Quantity 25 g
Formula Weight 391.53
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Trifluoromethanesulfonate