missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium Thiocyanate 97.0+%, TCI America™
Supplier: TCI America T1278250G
Specifications
| Tetrabutylammonium Thiocyanate | |
| 123°C | |
| C17H36N2S | |
| 250 g | |
| RSHBFZCIFFBTEW-UHFFFAOYSA-M | |
| tetrabutylazanium;thiocyanate | |
| 3014224 | |
| ≥97.0% (T) |
| 3674-54-2 | |
| White-Yellow | |
| MFCD00012140 | |
| Tetrabutylammonium Rhodanide | |
| CCCC[N+](CCCC)(CCCC)CCCC.C(#N)[S-] | |
| 300.549 | |
| 300.55 | |
| Crystalline Powder |
Chemical Identifiers
| 3674-54-2 | |
| 300.549 | |
| RSHBFZCIFFBTEW-UHFFFAOYSA-M | |
| 3014224 | |
| CCCC[N+](CCCC)(CCCC)CCCC.C(#N)[S-] |
| C17H36N2S | |
| MFCD00012140 | |
| Tetrabutylammonium Rhodanide | |
| tetrabutylazanium;thiocyanate |
Safety and Handling
TSCA : No