missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Thiocyanate 97.0+%, TCI America™

Catalog No. p-7136693
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T127825G 25 g
T1278250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T127825G Supplier TCI America Supplier No. T127825G
Only null left

Chemical Identifiers

CAS 3674-54-2
Molecular Formula C17H36N2S
Molecular Weight (g/mol) 300.549
MDL Number MFCD00012140
InChI Key RSHBFZCIFFBTEW-UHFFFAOYSA-M
Synonym Tetrabutylammonium Rhodanide
PubChem CID 3014224
IUPAC Name tetrabutylazanium;thiocyanate
SMILES CCCC[N+](CCCC)(CCCC)CCCC.C(#N)[S-]

Specifications

Melting Point 123°C
Color White-Yellow
Quantity 25 g
Formula Weight 300.55
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Thiocyanate