accessibility menu, dialog, popup

UserName

CAS RN 3674-54-2

IUPAC Name: cyanosulfanide; tetrabutylazanium
Synonyms: tetrabutylammonium ion thiocyanate
Molecular Weight (g/mol): 300.55
Molecular Formula: C17H36N2S
InChi Key: RSHBFZCIFFBTEW-UHFFFAOYSA-M
SMILES: [S-]C#N.CCCC[N+](CCCC)(CCCC)CCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results