missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Salicylate 98.0+%, TCI America™

Catalog No. T127625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T127625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T127625G Supplier TCI America Supplier No. T127625G
Only null left

Chemical Identifiers

CAS 22307-72-8
Molecular Formula C23H41NO3
Molecular Weight (g/mol) 379.585
MDL Number MFCD00145388
InChI Key ZWRWGKGPUFESNE-UHFFFAOYSA-M
PubChem CID 54707142
IUPAC Name 2-carboxyphenolate;tetrabutylazanium
SMILES CCCC[N+](CCCC)(CCCC)CCCC.C1=CC=C(C(=C1)C(=O)O)[O-]

Specifications

Melting Point 92°C
Color White
Quantity 25 g
Formula Weight 379.59
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Salicylate