missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Phosphate (0.5mol/L in Water) [Reagent for Ion-Pair Chromatography], TCI America™

Catalog No. I036710ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I036710ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I036710ML Supplier TCI America Supplier No. I036710ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5574-97-0
Molecular Formula C16H38NO4P
Molecular Weight (g/mol) 339.46
MDL Number MFCD00064526
InChI Key ARRNBPCNZJXHRJ-UHFFFAOYSA-M
Synonym tetrabutylammonium dihydrogen phosphate, tetrabutylammonium phosphate, tetrabutylammonium dihydrogenphosphate, 1-butanaminium, n,n,n-tributyl-, phosphate 1:1, tetra-n-butylammonium phosphate, tetrabutylazanium dihydrogen phosphate, dihydrogen phosphate; tetrabutylazanium, dihydrogen phosphate; tetrabutylammonium, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium ion dihydrogenphosphate ion
PubChem CID 2735142
IUPAC Name tetrabutylazanium dihydrogen phosphate
SMILES OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Color Colorless
pH 7.1 - 7.7
Quantity 10 mL
Physical Form Liquid
Chemical Name or Material Tetrabutylammonium Phosphate (0.5mol/L in Water) [Reagent for Ion-Pair Chromatography]
EINECSNumber (2)-0186
TSCA Yes