missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium p-Nitrophenoxide 98.0+%, TCI America™

Catalog No. T26695G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26695G 5 g
T266925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26695G Supplier TCI America Supplier No. T26695G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3002-48-0
Molecular Formula C22H40N2O3
Molecular Weight (g/mol) 380.573
InChI Key JWRHOHUAHHYNBL-UHFFFAOYSA-M
PubChem CID 18148
IUPAC Name 4-nitrophenolate;tetrabutylazanium
SMILES CCCC[N+](CCCC)(CCCC)CCCC.C1=CC(=CC=C1[N+](=O)[O-])[O-]

Specifications

Melting Point 145°C
Color Yellow
Quantity 5 g
Formula Weight 380.57
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium p-Nitrophenoxide