missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol for non-aq.titr.

Catalog No. AC123531000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC123531000 100 mL Glass bottle
AC123535000 500 mL Glass bottle
AC123530025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC123531000 Supplier Thermo Scientific Chemicals Supplier No. 123531000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol for non-aq.titr., Quantity: 500mL, Packaging: Glass bottle, Colorless to Yellow, Molecular Weight: 259.47, Beilstein: 04,II,634, CAS: 2052-49-5, 108-88-3, 67-56-1, Density: 0.8500g/mL, EINECS Number: 218-147-6 | CAS: 2052-49-5 | C16H37NO | 259.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst

Specifications

Chemical Name or Material Tetrabutylammonium hydroxide
CAS 2052-49-5, 108-88-3, 67-56-1
Color Colorless to Yellow
Density 0.8500g/mL
Flash Point 4°C
Packaging Glass bottle
Molecular Formula C16H37NO
Refractive Index 1.3775
Linear Formula [CH3(CH2)3]4NOH
MDL Number MFCD00009425
Quantity 100 mL
Beilstein 04,II,634
Fieser 05,645; 11,500; 07,332
Specific Gravity 0.85
Synonym tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
Solubility Information Solubility in water: insoluble
InChI Key VDZOOKBUILJEDG-UHFFFAOYSA-M
SMILES [OH-].CCCC[N+](CCCC)(CCCC)CCCC
IUPAC Name tetrabutylazanium;hydroxide
Molecular Weight (g/mol) 259.48
PubChem CID 2723671
Formula Weight 259.47
Physical Form Solution
Show More Show Less

RUO – Research Use Only