missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium hydrogen sulfate, 99%, extra pure

Catalog No. AC394205000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC394205000 500 g Glass bottle
AC394200050 5 g Glass bottle
AC394200250 25 g Glass bottle
AC394201000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC394205000 Supplier Thermo Scientific Chemicals Supplier No. 394205000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 32503-27-8 | C16H37NO4S | 339.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst

Chemical Identifiers

CAS 32503-27-8
Molecular Formula C16H37NO4S
Molecular Weight (g/mol) 339.54
MDL Number MFCD00011637
InChI Key SHFJWMWCIHQNCP-UHFFFAOYSA-M
Synonym tetrabutylammonium hydrogen sulfate, tetrabutylammonium hydrogensulfate, tetrabutylammonium bisulfate, tetra-n-butylammonium hydrogen sulfate, tbahs, tetrabutylammonium sulfate 1:1, 1-butanaminium, n,n,n-tributyl-, sulfate 1:1, tetrabutylammonium hydrogen sulphate, tetrabutylazanium hydrogen sulfate, ipc-tba-hs
PubChem CID 94433
IUPAC Name hydrogen sulfate;tetrabutylazanium
SMILES OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 168.0°C to 172.0°C
Color Brown-Yellow to Colorless
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula [CH3(CH2)3]4NHSO4
Quantity 500 g
Solubility Information (10% in water) Almost clear colorless
Formula Weight 339.53
Percent Purity 99%
Grade Extra Pure
Physical Form Solution
Chemical Name or Material Tetrabutylammonium hydrogen sulfate, Extra Pure
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 251-068-5

RUO – Research Use Only