missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Hydrogen Sulfate, 98%, Spectrum™ Chemical

Catalog No. 18614341
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614341 100 g Amber Glass Bottle
18614339 5 g Amber Glass Bottle
18614340 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18614341 Supplier Spectrum Chemical Mfg Cor Supplier No. T1294100GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 32503-27-8
Molecular Formula C16H37NO4S
Molecular Weight (g/mol) 339.54
InChI Key SHFJWMWCIHQNCP-UHFFFAOYSA-M
IUPAC Name tetrabutylazanium hydrogen sulfate
SMILES OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

CAS Max % 1
Melting Point 170° to 175°C
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 0.98
Grade Reagent
Physical Form Crystals or Flakes
Chemical Name or Material Tetrabutylammonium Hydrogen Sulfate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.