missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Tetrabutylammonium Chloride

Numéro de catalogue. 86870500G
Encompass
Modifier l'affichage
Click to view available options
Quantité:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
86870500G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Numéro de catalogue. 86870500G Fournisseur Sigma Aldrich Code fournisseur 86870500G
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Find details here.Used with phosphorus pentoxide for greener deoxychlorination.Process for Producing Halogenated Heteroaryl Compounds

Spécifications

CAS 1112-67-0
Formule moléculaire C16H36ClN
Formule linéaire [CH3(CH2)3]4NCl
Quantité 25 g
Numéro MDL MFCD00011635
Poids moléculaire (g/mol) 277.92 g/mol