missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromophthalic anhydride, 98%

Catalog No. AAB2455036
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2455036 500 g
AAB2455022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2455036 Supplier Thermo Scientific Chemicals Supplier No. B2455036
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 632-79-1 | C8Br4O3 | 463.701 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 632-79-1
Molecular Formula C8Br4O3
Molecular Weight (g/mol) 463.701
MDL Number MFCD00005919
InChI Key QHWKHLYUUZGSCW-UHFFFAOYSA-N
PubChem CID 12443
IUPAC Name 4,5,6,7-tetrabromo-2-benzofuran-1,3-dione
SMILES C12=C(C(=C(C(=C1Br)Br)Br)Br)C(=O)OC2=O

Specifications

Melting Point 274°C to 280°C
Density 2.87
Odor Odorless
Quantity 500 g
Beilstein 18908
Sensitivity Moisture sensitive
Synonym tetrabromophthalic anhydride, bromphthal, firemaster pht 4, phthalic anhydride, tetrabromo, bromophthal, tetrabromophthalic acid anhydride, firemaster pht4, 1,3-isobenzofurandione, 4,5,6,7-tetrabromo, 4,5,6,7-tetrabromo-1,3-isobenzofurandione, pht 4
Formula Weight 463.7
Percent Purity 98%
Chemical Name or Material Tetrabromophthalic anhydride
Hazard Category H315-H317-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
EINECSNumber 211-185-4
RTECSNumber TI3350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only