missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tetrabromophenolphthalein ethyl ester, pure

Catalog No. AC420070010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC420070010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC420070010 Supplier Thermo Scientific Chemicals Supplier No. 420070010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1176-74-5 | C22H14Br4O4 | 661.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1176-74-5
Molecular Formula C22H14Br4O4
Molecular Weight (g/mol) 661.96
MDL Number MFCD00066387
InChI Key SQFXATUXPUCFFO-UHFFFAOYSA-N
Synonym tetrabromophenolphthalein ethyl ester, 3',3,5',5-tetrabromophenolphthalein ethyl ester, ethyl tetrabromophenolphthalein, unii-6d91ns781o, ethyl 2-3,5-dibromo-4-hydroxyphenyl 3,5-dibromo-4-oxo-2,5-cyclohexadien-1-ylidene methyl benzoate, 3 inverted exclamation marka,3 inverted exclamation marka,5 inverted exclamation marka,5 inverted exclamation marka-tetrabromophenolphthalein ethyl ester, benzoic acid, 2-3,5-dibromo-4-hydroxyphenyl 3,5-dibromo-4-oxo-2,5-cyclohexadien-1-ylidene methyl-, ethyl ester, benzoic acid,2-3,5-dibromo-4-hydroxyphenyl 3,5-dibromo-4-oxo-2,5-cyclohexadien-1-ylidene methyl-,ethyl ester, ethyl 2-3,5-dibromo-4-hydroxyphenyl 3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene methyl benzoate
PubChem CID 92873
IUPAC Name ethyl 2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzoate
SMILES CCOC(=O)C1=CC=CC=C1C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br

Specifications

Melting Point 208.0°C to 211.0°C
Color Red to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 9331
Solubility Information Solubility in water: negligible.
Spectrophotometry 80.0% min.
Formula Weight 661.96
Grade Pure
Physical Form Powder
Chemical Name or Material Tetrabromophenolphthalein ethyl ester
Show More Show Less
EINECSNumber 214-645-2

RUO – Research Use Only