missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromohydroquinone 97.0+%, TCI America™

Catalog No. T17905G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17905G 5 g
T179025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17905G Supplier TCI America Supplier No. T17905G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2641-89-6
Molecular Formula C6H2Br4O2
Molecular Weight (g/mol) 425.696
MDL Number MFCD00013970
InChI Key DTFQULSULHRJOA-UHFFFAOYSA-N
Synonym tetrabromohydroquinone, 1,4-dihydroxytetrabromobenzene, tetrabromoquinol, 2,3,5,6-tetrabromohydroquinone, 2,3,5,6-tetrabromo-1,4-benzenediol, 1,2-benzenediol,3,4,5,6-tetrabromo, 1,4-benzenediol, 2,3,5,6-tetrabromo, 1,2,4,5-tetrabromo-3,6-dihydroxybenzene
PubChem CID 75840
IUPAC Name 2,3,5,6-tetrabromobenzene-1,4-diol
SMILES C1(=C(C(=C(C(=C1Br)Br)O)Br)Br)O

Specifications

Quantity 5 g
Formula Weight 425.70
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabromohydroquinone