missing translation for 'onlineSavingsMsg'
Learn More

Tetrabenzyl pyrophosphate, 98%

Catalog No. AAL0908303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0908303 1 g
AAL0908306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0908303 Supplier Thermo Scientific Chemicals Supplier No. L0908303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 990-91-0 | C28H28O7P2 | 538.473 g/mol

Tetrabenzyl pyrophosphate is used in the preparation of phosphoryl derivatives of shikimic acid in the presence of LDA. It is used in the preparation of dibenzyl phosphoro fluoridate in the presence of cesium fluoride as a catalyst. It is also used as a precursor in pharmaceuticals and involved in the phosphorylation of inositol derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrabenzyl pyrophosphate is used in the preparation of phosphoryl derivatives of shikimic acid in the presence of LDA. It is used in the preparation of dibenzyl phosphoro fluoridate in the presence of cesium fluoride as a catalyst. It is also used as a precursor in pharmaceuticals and involved in the phosphorylation of inositol derivatives.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acid and bases and reducing agents.

Chemical Identifiers

CAS 990-91-0
Molecular Formula C28H28O7P2
Molecular Weight (g/mol) 538.473
MDL Number MFCD00051941
InChI Key NSBNXCZCLRBQTA-UHFFFAOYSA-N
Synonym tetrabenzyl pyrophosphate, tetrabenzyl diphosphate, pyrophosphoric acid tetrabenzyl ester, tetrabenzylpyrophosphate, diphosphoric acid, tetrakis phenylmethyl ester, dibenzyl dibenzyloxyphosphoryl oxyphosphonate, benzyl pyrophosphate, acmc-209sbp, tetra benzyl pyrophosphate, tetrabenzyl diphosphate #
PubChem CID 563183
IUPAC Name dibenzyl bis(phenylmethoxy)phosphoryl phosphate
SMILES C1=CC=C(C=C1)COP(=O)(OCC2=CC=CC=C2)OP(=O)(OCC3=CC=CC=C3)OCC4=CC=CC=C4

Specifications

Melting Point 58°C to 64°C
Quantity 1 g
UN Number UN3261
Beilstein 2068292
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 538.48
Percent Purity 98%
Chemical Name or Material Tetrabenzyl pyrophosphate
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA No
Recommended Storage Store at -20°C; Store under Argon

RUO – Research Use Only