missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylphosphonium bromide, 99%

Catalog No. AAA1086836
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1086836 500 g
AAA1086822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1086836 Supplier Thermo Scientific Chemicals Supplier No. A1086836
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3115-68-2 | C16H36BrP | 339.342 g/mol

Tetra-n-butylphosphonium bromide is used as a phase-transfer catalyst at high temperatures, at which nitrogen analogues tend to undergo elimination e.g. as catalyst for the dealkoxycarbonylation of malonates and β-keto esters by heating in stearic acid. used as an ionic liquid solvent in the regioselective O-alkylation of C/O ambident nucleophiles. It is a synthetic reagent used in proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-butylphosphonium bromide is used as a phase-transfer catalyst at high temperatures, at which nitrogen analogues tend to undergo elimination e.g. as catalyst for the dealkoxycarbonylation of malonates and ß-keto esters by heating in stearic acid. used as an ionic liquid solvent in the regioselective O-alkylation of C/O ambident nucleophiles. It is a synthetic reagent used in proteomics research.

Solubility
Soluble in water (70 g/100 ml)

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature.

Chemical Identifiers

CAS 3115-68-2
Molecular Formula C16H36BrP
Molecular Weight (g/mol) 339.342
MDL Number MFCD00011853
InChI Key RKHXQBLJXBGEKF-UHFFFAOYSA-M
Synonym tetrabutylphosphonium bromide, tetra-n-butylphosphonium bromide, phosphonium, tetrabutyl-, bromide, px 4b, tetra butyl phosphonium bromide, tetrabutylphosphoniumbromide, tetrabutylphosphanium bromide, tetrabutyl phosphonium bromide, phosphonium, tetrabutyl-, bromide 1:1, acmc-1clh3
PubChem CID 76564
IUPAC Name tetrabutylphosphanium;bromide
SMILES CCCC[P+](CCCC)(CCCC)CCCC.[Br-]

Specifications

Melting Point 99°C to 103°C
Flash Point 290°C (554°F)
Linear Formula [CH3(CH2)3]4PBr
Quantity 500 g
UN Number UN3464
Beilstein 4160474
Sensitivity Hygroscopic
Solubility Information Soluble in water (70g/100 ml)
Formula Weight 339.35
Percent Purity 99%
Chemical Name or Material Tetra-n-butylphosphonium bromide
Show More Show Less
Hazard Category H302+H332-H311-H315-H319-H335
Hazard Statement GHS H Statement
H310-H318-H302-H332-H315-H335
Fatal in contact with skin.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.
EINECSNumber 221-487-8
RTECSNumber TA2417000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only