missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium fluoride, 75% w/w aq. soln.

Catalog No. AAB2126136
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2126136 500 g
AAB2126114 25 g
AAB2126122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2126136 Supplier Thermo Scientific Chemicals Supplier No. B2126136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 429-41-4 | C16H36FN | 261.47 g/mol

Tetrabutylammonium Fluoride is a reactant used for the synthesis of multiple compounds, including conjugated dienoic acid esters, oligoribonucleotides with phosphonate-modified linkages, and triple monoamine reuptake inhibitors, among others. It has proven useful in studying cancer, diabetes, and neurochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrabutylammonium Fluoride is a reactant used for the synthesis of multiple compounds, including conjugated dienoic acid esters, oligoribonucleotides with phosphonate-modified linkages, and triple monoamine reuptake inhibitors, among others. It has proven useful in studying cancer, diabetes, and neurochemistry.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.Keep away from strong oxidizing agents,

Chemical Identifiers

CAS 429-41-4
Molecular Formula C16H36FN
Molecular Weight (g/mol) 261.47
MDL Number MFCD00011747
InChI Key FPGGTKZVZWFYPV-UHFFFAOYSA-M
Synonym tetrabutylammonium fluoride, tbaf, tetrabutylazanium fluoride, tetrabutyl ammonium fluoride, tetra-n-butylammonium fluoride, tetrabutylamine, fluoride, n,n,n-tributylbutan-1-aminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, n,n,n-tributyl-1-butanaminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride 1:1
PubChem CID 2724141
ChEBI CHEBI:51990
IUPAC Name tetrabutylazanium;fluoride
SMILES [F-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Quantity 500 g
UN Number UN2922
Solubility Information Insoluble in water.
Formula Weight 261.47
Concentration or Composition (by Analyte or Components) 75% w/w aq. soln.
Chemical Name or Material Tetra-n-butylammonium fluoride
Hazard Category H301-H314-H335
Hazard Statement GHS H Statement
H300-H314-H318
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 207-057-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only