Learn More
LaMotte™ Test Strips
For chlorine, iodine and QAC testing
$27.38 - $35.35
Specifications
For Use With (Application) Test drinking water, industrial water for boiling and cooling, laundry or sanitation water and wastewater, for chlorine, iodine and QAC testing
View More Specs
Products 5
Catalog Number Mfr. No. Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Type Quantity Price Quantity & Availability  
22307864
View Documents Promotion Details
LaMotte™
2951
QAC Test Strip 100 Pk.
Pack of 100 Each for $27.38
Only null left
03920385
View Documents Promotion Details
LaMotte™
2951HR
High Range QAC Test Paper Each
Each for $35.35
Only null left
22479808
View Documents Promotion Details
LaMotte™
4250BJ
Chlorine Test Paper 200 Pk.
Pack of 200 Each for $28.03
Only null left
22307863
View Documents Promotion Details
LaMotte™
2948BJ
Iodine Test Strip 200 Pk.
Pack of 200 Each for $28.53
Only null left
03920453
View Documents Promotion Details
LaMotte™
3072J
QAC Test Strip 100 Strips
Each for $28.74
Only null left
Description
Description
Test chlorine from 10 to 200ppm, iodine from 12 to 100ppm and quaternary ammonium compounds from 50-400ppm, or from 200 to 1500ppm
  • Chemically treated paper strips produce color changes in as little as 5 seconds
  • Simply dip strip into sample
  • Compare strip color to color chart located on plastic vial
  • Plastic, waterproof vials are labeled with test instructions
  • Desiccant cap
Test drinking water, industrial water for boiling and cooling, laundry /sanitation water and wastewater
Order Info
Iodine and chlorine papers - packed 200 per plastic vial; QAC strips - packed in plastic vials of 100; Desiccant cap for longer life
Specifications
Specifications
Test drinking water, industrial water for boiling and cooling, laundry or sanitation water and wastewater, for chlorine, iodine and QAC testing
Videos
Safety and Handling
Safety and Handling
ShelfLife : 1 Year