Learn More
LaMotte™ Test Strips
Description
Test chlorine from 10 to 200ppm, iodine from 12 to 100ppm and quaternary ammonium compounds from 50-400ppm, or from 200 to 1500ppm
- Chemically treated paper strips produce color changes in as little as 5 seconds
- Simply dip strip into sample
- Compare strip color to color chart located on plastic vial
- Plastic, waterproof vials are labeled with test instructions
- Desiccant cap
Test drinking water, industrial water for boiling and cooling, laundry /sanitation water and wastewater
Order Info
Iodine and chlorine papers - packed 200 per plastic vial; QAC strips - packed in plastic vials of 100; Desiccant cap for longer life
Spécifications
Spécifications
| For Use With (Application) | Test drinking water, industrial water for boiling and cooling, laundry or sanitation water and wastewater, for chlorine, iodine and QAC testing |
| Includes | Color chart, plastic waterproof opaque vial, desiccant cap |
| Quantity | 100 Pk. |
| Range | 50-100-200-400 ppm |
| Type | QAC Test Strip |