Learn More
LaMotte™ Test Strips
For chlorine, iodine and QAC testing
Supplier: LaMotte™ 4250BJ
Description
Test chlorine from 10 to 200ppm, iodine from 12 to 100ppm and quaternary ammonium compounds from 50-400ppm, or from 200 to 1500ppm
- Chemically treated paper strips produce color changes in as little as 5 seconds
- Simply dip strip into sample
- Compare strip color to color chart located on plastic vial
- Plastic, waterproof vials are labeled with test instructions
- Desiccant cap
Test drinking water, industrial water for boiling and cooling, laundry /sanitation water and wastewater
Order Info
Iodine and chlorine papers - packed 200 per plastic vial; QAC strips - packed in plastic vials of 100; Desiccant cap for longer life
Chemical Identifiers
| Test drinking water, industrial water for boiling and cooling, laundry or sanitation water and wastewater, for chlorine, iodine and QAC testing |
Specifications
| Test drinking water, industrial water for boiling and cooling, laundry or sanitation water and wastewater, for chlorine, iodine and QAC testing | |
| 200 Pk. | |
| Chlorine Test Paper |
| Color chart, plastic vial | |
| 10-50-100-200 ppm |
Safety and Handling
ShelfLife : 1 Year