Learn More
LaMotte™ Test Strips
For chlorine, iodine and QAC testing
Supplier:  LaMotte™ 2951HR
 View more versions of this product
Catalog No. 03920385
Only null left
Description
Description
Test chlorine from 10 to 200ppm, iodine from 12 to 100ppm and quaternary ammonium compounds from 50-400ppm, or from 200 to 1500ppm
  • Chemically treated paper strips produce color changes in as little as 5 seconds
  • Simply dip strip into sample
  • Compare strip color to color chart located on plastic vial
  • Plastic, waterproof vials are labeled with test instructions
  • Desiccant cap
Test drinking water, industrial water for boiling and cooling, laundry /sanitation water and wastewater
Order Info
Iodine and chlorine papers - packed 200 per plastic vial; QAC strips - packed in plastic vials of 100; Desiccant cap for longer life
Specifications
Chemical Identifiers
Test drinking water, industrial water for boiling and cooling, laundry or sanitation water and wastewater, for chlorine, iodine and QAC testing
Specifications
Test drinking water, industrial water for boiling and cooling, laundry or sanitation water and wastewater, for chlorine, iodine and QAC testing
Each
High Range QAC Test Paper
Color chart, plastic vial
200-400-600-1000-1500 ppm
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
ShelfLife : 1 Year