missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylhydroquinone 98.0+%, TCI America™

Catalog No. B083325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B083325G 25 g
B0833500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B083325G Supplier TCI America Supplier No. B083325G

Chemical Identifiers

CAS 1948-33-0
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00002344
InChI Key BGNXCDMCOKJUMV-UHFFFAOYSA-N
Synonym tert-butylhydroquinone, tbhq, 2-tert-butylhydroquinone, t-butylhydroquinone, mtbhq, t-butyl hydroquinone, 2-tert-butyl-1,4-benzenediol, 2-t-butylhydroquinone, mono-tert-butylhydroquinone, sustane
PubChem CID 16043
ChEBI CHEBI:78886
IUPAC Name 2-tert-butylbenzene-1,4-diol
SMILES CC(C)(C)C1=C(C=CC(=C1)O)O

Specifications

Melting Point 127°C
Boiling Point 288°C
Quantity 25 g
UN Number 3077
Formula Weight 166.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butylhydroquinone
EINECSNumber (3)-0549
RTECSNumber MX4375000
TSCA Yes