missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyldimethylsilyl N-Phenylbenzimidate 98.0+%, TCI America™
Supplier: TCI America B26975G
Specifications
| tert-Butyldimethylsilyl N-Phenylbenzimidate | |
| 71°C | |
| C19H25NOSi | |
| 5 g | |
| CQKWSHDUCPVOAI-UHFFFAOYSA-N | |
| tert-butyldimethylsilyl N-phenylbenzenecarboximidate | |
| 11197712 | |
| ≥98.0% (HPLC) |
| 404392-70-7 | |
| White | |
| MFCD08276306 | |
| O-(tert-Butyldimethylsilyl)benzanilide, TBDMS-BEZA | |
| CC(C)(C)[Si](C)(C)OC(=NC1=CC=CC=C1)C1=CC=CC=C1 | |
| 311.50 | |
| 311.50 | |
| Crystalline Powder |
Chemical Identifiers
| 404392-70-7 | |
| 311.50 | |
| CQKWSHDUCPVOAI-UHFFFAOYSA-N | |
| 11197712 | |
| CC(C)(C)[Si](C)(C)OC(=NC1=CC=CC=C1)C1=CC=CC=C1 |
| C19H25NOSi | |
| MFCD08276306 | |
| O-(tert-Butyldimethylsilyl)benzanilide, TBDMS-BEZA | |
| tert-butyldimethylsilyl N-phenylbenzenecarboximidate |
Safety and Handling
TSCA : No