missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyldimethylchlorosilane, 50% w/w in toluene, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 50 g |
| Flash Point | 7°C (45°F) |
| Odor | Characteristic |
| Sensitivity | Moisture sensitive |
| Density | 0.87 |
| Linear Formula | (CH3)3CSi(CH3)2Cl |
| UN Number | UN2924 |
| Formula Weight | 150.73 |
| Concentration or Composition (by Analyte or Components) | 50% w/w in toluene |
| Chemical Name or Material | tert-Butyldimethylchlorosilane |
Safety and Handling
| Hazard Category | H225-H304-H314-H335-H336-H361d-H373 |
| Hazard Statement | GHS H Statement H225-H314-H318-H361-H373-H336 Highly flammable liquid and vapour. Causes severe skin burns and eye damage. Causes serious eye damage. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. May cause drowsiness or dizziness. |
| Precautionary Statement | P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S. |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only